4-(2-Bromphenylcarbamoyl)phenylboronsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-((2-Bromophenyl)carbamoyl)phenyl)boronic acid

OR3498-1G
APOSOR3498-1GEA 21.9 EUR
APOSOR3498-1G
4-(2-Bromphenylcarbamoyl)phenylboronsäure 95%
4-(2-Bromphenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BBrNO₃
Molecular Weight: 319,9500 g/mol
Schmelzpunkt: 233…235 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027202
CAS-Nummer: 874288-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR