4'-Amino-3',5'-dichloracetophenon
Lieferant: Apollo Scientific
Synonyme:
4′-Amino-3′,5′-dichloroacetophenone
OR350012-100G
APOSOR350012-100GEA
95.7
EUR
APOSOR350012-100G
4'-Amino-3',5'-dichloracetophenon
4'-Amino-3',5'-dichloracetophenon
Formel:
C₈H₇Cl₂NO Molecular Weight: 204,0500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00238535 CAS-Nummer: 37148-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...