cis-4-Aminocyclohexanol

Lieferant: Apollo Scientific

OR350015-1G
APOSOR350015-1GEA 98.6 EUR
APOSOR350015-1G
cis-4-Aminocyclohexanol
cis-4-Aminocyclohexanol
Formel: C₆H₁₃NO
Molecular Weight: 115,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09997786
CAS-Nummer: 40525-78-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR