Benzyl (1-phenylcyclopropyl)carbamate

Lieferant: Apollo Scientific

OR350025-1G
APOSOR350025-1GEA 58 EUR
APOSOR350025-1G
Benzyl (1-phenylcyclopropyl)carbamate
Benzyl (1-phenylcyclopropyl)carbamate
Formel: C₁₇H₁₇NO₂
Molecular Weight: 267,322 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205767
CAS-Nummer: 1324000-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR