1-Benzyl-4-(Boc-amino)piperidine

Lieferant: Apollo Scientific

OR350038-25G
APOSOR350038-25GEA 33 EUR
APOSOR350038-25G
1-Benzyl-4-(Boc-amino)piperidine
1-Benzyl-4-(Boc-amino)piperidine
Formel: C₁₇H₂₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03093979
CAS-Nummer: 73889-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR