6-Bromo[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one

OR350046-1G
APOSOR350046-1GEA 130 EUR
APOSOR350046-1G
6-Bromo[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one
6-Bromo[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one
Formel: C₆H₄BrN₃O
Molecular Weight: 214,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11110678
CAS-Nummer: 425702-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR