Benzo[d][1,3]dioxol-4-amine

Lieferant: Apollo Scientific
Achtung

OR350054-1G
APOSOR350054-1GEA 66.7 EUR
APOSOR350054-1G
Benzo[d][1,3]dioxol-4-amine
Benzo[d][1,3]dioxol-4-amine
Formel: C₇H₇NO₂
Molecular Weight: 137.1400 g/mol
Schmelzpunkt: 136…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09038121
CAS-Nummer: 1668-84-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR