1-(tert-Butyl)-1H-pyrazol-5-amine

Lieferant: Apollo Scientific

OR350060-1G
APOSOR350060-1GEA 125 EUR
APOSOR350060-1G
1-(tert-Butyl)-1H-pyrazol-5-amine
1-(tert-Butyl)-1H-pyrazol-5-amine
Formel: C₇H₁₃N₃
Molecular Weight: 139,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11054260
CAS-Nummer: 442850-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR