(4-Bromphenoxy)essigsäure

Lieferant: Apollo Scientific

Synonyme: 4-Bromophenoxyacetic acid

OR350064-100G
APOSOR350064-100GEA 69.6 EUR
APOSOR350064-100G
(4-Bromphenoxy)essigsäure
(4-Bromphenoxy)essigsäure
Formel: (C₆H₄Br)OCH₂COOH
Molecular Weight: 231,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00093070
CAS-Nummer: 1878-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR