1-Boc-4-(tosyloxy)piperidin

Lieferant: Apollo Scientific

OR350072-1G
APOSOR350072-1GEA 0 EUR
APOSOR350072-1G
1-Boc-4-(tosyloxy)piperidin
1-Boc-4-(tosyloxy)piperidin
Formel: C₁₇H₂₅NO₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD06796535
CAS-Nummer: 118811-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR