2'-Chloropropiophenone
Lieferant: Apollo Scientific
Synonyme:
2-Chlorophenyl Ethyl Ketone, 1-(2-Chlorophenyl)propan-1-one
OR350082-1G
APOSOR350082-1GEA
62.5
EUR
APOSOR350082-1G
2'-Chloropropiophenone
2'-Chloropropiophenone
Formel:
C₉H₉ClO Molecular Weight: 168,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07787256 CAS-Nummer: 6323-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...