(2-Chloro-4-isopropoxyphenyl)boronic acid

Lieferant: Apollo Scientific
Achtung

OR350084-250MG
APOSOR350084-250MGEA 21.9 EUR
APOSOR350084-250MG
(2-Chloro-4-isopropoxyphenyl)boronic acid
(2-Chloro-4-isopropoxyphenyl)boronic acid
Formel: C₉H₁₂BClO₃
Molecular Weight: 214,454 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02684319
CAS-Nummer: 313545-47-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR