4-Cyano-2-methoxybenzeneboronic acid
Lieferant: Apollo Scientific
Synonyme:
(4-Cyano-2-methoxyphenyl)boronic acid
OR350088-1G
APOSOR350088-1GEA
75.4
EUR
APOSOR350088-1G
4-Cyano-2-methoxybenzeneboronic acid
4-Cyano-2-methoxybenzeneboronic acid
Formel:
C₈H₈BNO₃ Molecular Weight: 176,966 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16618929 CAS-Nummer: 1256345-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...