1-(2-Chloropyridin-4-yl)piperazine

Lieferant: Apollo Scientific

OR350092-1G
APOSOR350092-1GEA 0 EUR
APOSOR350092-1G
1-(2-Chloropyridin-4-yl)piperazine
1-(2-Chloropyridin-4-yl)piperazine
Formel: C₉H₁₂ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06738480
CAS-Nummer: 854159-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR