2-Chloro-4,4'-bipyridine

Lieferant: Apollo Scientific

OR350095-250MG
APOSOR350095-250MGEA 101 EUR
APOSOR350095-250MG
2-Chloro-4,4'-bipyridine
2-Chloro-4,4'-bipyridine
Formel: C₁₀H₇ClN₂
Molecular Weight: 190,629 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11111098
CAS-Nummer: 53344-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR