1-(4-Chloropyridin-2-yl)piperazine
Lieferant: Apollo Scientific
OR350103-1G
APOSOR350103-1GEA
119
EUR
APOSOR350103-1G
1-(4-Chloropyridin-2-yl)piperazine
1-(4-Chloropyridin-2-yl)piperazine
Formel:
C₉H₁₂ClN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06738950 CAS-Nummer: 885277-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...