4-Chloro-N-methylpyrimidin-2-amine

Lieferant: Apollo Scientific

OR350105-1G
APOSOR350105-1GEA 219 EUR
APOSOR350105-1G
4-Chloro-N-methylpyrimidin-2-amine
4-Chloro-N-methylpyrimidin-2-amine
Formel: C₅H₆ClN₃
Molecular Weight: 143,5700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09834973
CAS-Nummer: 22404-46-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR