4-(Dimethylamino)zimtsäure
Lieferant: Apollo Scientific
OR350131-5G
APOSOR350131-5GEA
0
EUR
APOSOR350131-5G
4-(Dimethylamino)zimtsäure
4-(Dimethylamino)zimtsäure
Formel:
C₁₁H₁₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004397 CAS-Nummer: 1552-96-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...