2,4-Di-tert-butyl-5-nitrophenol 98%
Lieferant: Apollo Scientific
OR350138-1G
APOSOR350138-1GEA
109
EUR
APOSOR350138-1G
2,4-Di-tert-butyl-5-nitrophenol 98%
2,4-Di-tert-butyl-5-nitrophenol
Formel:
C₁₄H₂₁NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11052606 CAS-Nummer: 873055-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...