4-oxo-3,4-Dihydroquinazoline-6-carbonitrile

Lieferant: Apollo Scientific

OR350144-250MG
APOSOR350144-250MGEA 178 EUR
APOSOR350144-250MG
4-oxo-3,4-Dihydroquinazoline-6-carbonitrile
4-oxo-3,4-Dihydroquinazoline-6-carbonitrile
Formel: C₉H₅N₃O
Molecular Weight: 171,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20039986
CAS-Nummer: 117297-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR