3,5-Dimethylbenzonitril
Lieferant: Apollo Scientific
OR350145-25G
APOSOR350145-25GEA
59.6
EUR
APOSOR350145-25G
3,5-Dimethylbenzonitril
3,5-Dimethylbenzonitril
Formel:
C₉H₉N Molecular Weight: 131,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060670 CAS-Nummer: 22445-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...