5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-3-carboxylic acid

Lieferant: Apollo Scientific

OR350151-1G
APOSOR350151-1GEA 177 EUR
APOSOR350151-1G
5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-3-carboxylic acid
5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-3-carboxylic acid
Formel: C₇H₈N₂O₂
Molecular Weight: 152,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11846937
CAS-Nummer: 796729-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR