Ethylcyclobutancarboxylat
Lieferant: Apollo Scientific
OR350159-100G
APOSOR350159-100GEA
140
EUR
APOSOR350159-100G
Ethylcyclobutancarboxylat
Ethylcyclobutancarboxylat
Formel:
C₇H₁₂O₂ Molecular Weight: 128,1690 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001321 CAS-Nummer: 14924-53-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...