trans-4-Ethylcyclohexancarbonsäure

Lieferant: Apollo Scientific

OR350166-100G
APOSOR350166-100GEA 84.1 EUR
APOSOR350166-100G
trans-4-Ethylcyclohexancarbonsäure
trans-4-Ethylcyclohexanecarboxylic acid
Formel: C₉H₁₆O₂
Molecular Weight: 156,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01076323
CAS-Nummer: 6833-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR