Mecarbinat
Lieferant: Apollo Scientific
Synonyme:
Ethyl 5-Hydroxy-1,2-dimethylindole-3-carboxylate, 5-Hydroxy-1,2-dimethylindole-3-carboxylic Acid Ethyl Ester
OR350169-5G
APOSOR350169-5GEA
125
EUR
APOSOR350169-5G
Mecarbinat
Mecarbinat
Formel:
C₁₃H₁₅NO₃ Molecular Weight: 233,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00451373 CAS-Nummer: 15574-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...