Ethyl-4-methylpyrrol-2-carboxylat 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 4-Methyl-2-pyrrolecarboxylate
OR350171-1G
APOSOR350171-1GEA
154
EUR
APOSOR350171-1G
Ethyl-4-methylpyrrol-2-carboxylat 95%
Ethyl-4-methylpyrrol-2-carboxylat
Formel:
C₈H₁₁NO₂ Molecular Weight: 153,1800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191668 CAS-Nummer: 40611-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...