Ethyl-4-cyanbenzoat
Lieferant: Apollo Scientific
Synonyme:
4-Cyanbenzoesäureethylester
OR350177-25G
APOSOR350177-25GEA
72.5
EUR
APOSOR350177-25G
Ethyl-4-cyanbenzoat
Ethyl-4-cyanbenzoat
Formel:
C₁₀H₉NO₂ Molecular Weight: 175.19 g/mol Siedepunkt: 152 Schmelzpunkt: 54 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001824 CAS-Nummer: 7153-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...