N-alpha-FMOC-Nepsilon-BOC-L-Lysin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-ε-(tert-Butoxycarbonyl)-N-ɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-((tert-butoxycarbonyl)amino)hexanoic acid, N-ε-Boc-N-ɑ-Fmoc-L-lysine, Fmoc-Lys(Boc)-OH

OR350184-100G OR350184-25G OR350184-500G
APOSOR350184-100GEA 75.4 EUR
APOSOR350184-100G APOSOR350184-25G APOSOR350184-500G
N-alpha-FMOC-Nepsilon-BOC-L-Lysin 97%
N-alpha-FMOC-Nepsilon-BOC-L-Lysin
Formel: C₂₆H₃₂N₂O₆
Schmelzpunkt: 129…135 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00037138
CAS-Nummer: 71989-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR