N-alpha-FMOC-Nepsilon-BOC-L-Lysin 97%

Lieferant: Apollo Scientific
Achtung

OR350184-100G OR350184-25G OR350184-500G
APOSOR350184-100GEA 0 EUR
APOSOR350184-100G APOSOR350184-25G APOSOR350184-500G
N-alpha-FMOC-Nepsilon-BOC-L-Lysin 97%
N-alpha-FMOC-Nepsilon-BOC-L-Lysin
Formel: C₂₆H₃₂N₂O₆
Schmelzpunkt: 129…135 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00037138
CAS-Nummer: 71989-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR