Hexadecyl 3,5-Di-tert-butyl-4-hydroxybenzoate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Hexadecyl 3,5-di-tert-butyl-4-hydroxybenzoate, 3,5-Di-tert-butyl-4-hydroxybenzoic acid Hexadecyl ester

OR350187-25G
APOSOR350187-25GEA 21.9 EUR
APOSOR350187-25G
Hexadecyl 3,5-Di-tert-butyl-4-hydroxybenzoate 98%
Hexadecyl 3,5-Di-tert-butyl-4-hydroxybenzoate
Formel: C₃₁H₅₄O₃
Molecular Weight: 474,77 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00180757
CAS-Nummer: 67845-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR