(3-(1-Hydroxyethyl)phenyl)boronic acid

Lieferant: Apollo Scientific

OR350193-1G
APOSOR350193-1GEA 87 EUR
APOSOR350193-1G
(3-(1-Hydroxyethyl)phenyl)boronic acid
(3-(1-Hydroxyethyl)phenyl)boronic acid
Formel: C₈H₁₁BO₃
Molecular Weight: 165,982 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112541
CAS-Nummer: 1036760-03-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR