2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)acetic acid
Lieferant: Apollo Scientific
OR350194-1G
APOSOR350194-1GEA
224
EUR
APOSOR350194-1G
2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)acetic acid
2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)acetic acid
Formel:
C₉H₈N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10698203 CAS-Nummer: 1912-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...