3-Hydroxy-4-methoxybenzylalkohol

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxy-4-(hydroxymethyl)anisol, 5-(Hydroxymethyl)-2-methoxyphenol, (3-Hydroxy-4-methoxyphenyl)methanol

OR350196-100G
APOSOR350196-100GEA 115 EUR
APOSOR350196-100G
3-Hydroxy-4-methoxybenzylalkohol
3-Hydroxy-4-methoxybenzylalkohol
Formel: C₈H₁₀O₃
Molecular Weight: 154,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004644
CAS-Nummer: 4383-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR