(3-(2-Hydroxypropan-2-yl)phenyl)boronic acid

Lieferant: Apollo Scientific

OR350197-1G
APOSOR350197-1GEA 162 EUR
APOSOR350197-1G
(3-(2-Hydroxypropan-2-yl)phenyl)boronic acid
(3-(2-Hydroxypropan-2-yl)phenyl)boronic acid
Formel: C₉H₁₃BO₃
Molecular Weight: 180,009 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09992906
CAS-Nummer: 955369-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR