(4-(2-Hydroxypropan-2-yl)phenyl)boronic acid

Lieferant: Apollo Scientific
Achtung

OR350200-1G
APOSOR350200-1GEA 60.9 EUR
APOSOR350200-1G
(4-(2-Hydroxypropan-2-yl)phenyl)boronic acid
(4-(2-Hydroxypropan-2-yl)phenyl)boronic acid
Formel: C₉H₁₃BO₃
Molecular Weight: 180,009 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09992905
CAS-Nummer: 886593-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR