5-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole

Lieferant: Apollo Scientific

OR350206-1G
APOSOR350206-1GEA 62.5 EUR
APOSOR350206-1G
5-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole
5-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole
Formel: C₈H₁₁IN₂O
Molecular Weight: 278,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19237232
CAS-Nummer: 1311197-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR