(2S)-Amino(cyclohexyl)essigsäure
Lieferant: Apollo Scientific
Synonyme:
H-Chg-OH, H-Cyclohexyl-Gly-OH, (S)-alpha-Aminocyclohexylacetic Acid
OR350213-100G
APOSOR350213-100GEA
63.8
EUR
APOSOR350213-100G
(2S)-Amino(cyclohexyl)essigsäure
(S)-2-Amino-2-cyclohexylacetic acid
Formel:
C₈H₁₅NO₂ Molecular Weight: 157,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01311679 CAS-Nummer: 14328-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...