Methylcyclohexancarboxylat > 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR350217-25G
APOSOR350217-25GEA
21.9
EUR
APOSOR350217-25G
Methylcyclohexancarboxylat > 98%
Methylcyclohexancarboxylat
Formel:
C₈H₁₄O₂ Molecular Weight: 142.2 g/mol Siedepunkt: 183 °C Dichte: 0.98 g/cm³ (25 °C) Flammpunkt: 60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001458 CAS-Nummer: 4630-82-4 UN: 3272 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...