3-Formyl-2-picolin 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Methylnicotinaldehyde

OR350218-1G OR350218-5G
APOSOR350218-1GEA 63.8 EUR
APOSOR350218-1G APOSOR350218-5G
3-Formyl-2-picolin 97%
3-Formyl-2-picolin
Formel: C₇H₇NO
Molecular Weight: 121.14 g/mol
Dichte: 1.106 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD08234911
CAS-Nummer: 60032-57-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR