1-Methyl-1H-pyrrol-2-carbaldehyd
Lieferant: Apollo Scientific
Synonyme:
N-Methylpyrrole-2-carboxaldehyde, 1-Methylpyrrole-2-aldehyde, 1-Methylpyrrole-2-carboxaldehyde, 1-Methylpyrrol-2-carbaldehyde, 2-Formyl-1-methylpyrrole
OR350219-100G
APOSOR350219-100GEA
163
EUR
APOSOR350219-100G
1-Methyl-1H-pyrrol-2-carbaldehyd
1-Methyl-2-pyrrolecarboxaldehyde
Formel:
C₆H₇NO Molecular Weight: 109,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003087 CAS-Nummer: 1192-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...