Methyl-3-piperidincarboxylat

Lieferant: Apollo Scientific

Synonyme: Methyl piperidine-3-carboxylate

OR350220-100G
APOSOR350220-100GEA 104 EUR
APOSOR350220-100G
Methyl-3-piperidincarboxylat
Methyl-3-piperidincarboxylat
Formel: C₇H₁₃NO₂
Molecular Weight: 143,1836 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00797850
CAS-Nummer: 50585-89-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR