3,5-Dihydroxytoluol Monohydrat
Lieferant: Apollo Scientific
Synonyme:
Orcin Monohydrat, 5-Methylresorcin Monohydrat
OR350224-100G
APOSOR350224-100GEA
209
EUR
APOSOR350224-100G
3,5-Dihydroxytoluol Monohydrat
3,5-Dihydroxytoluol Monohydrat
Formel:
C₇H₈O₂·1H₂O Molecular Weight: 142,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149092 CAS-Nummer: 6153-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...