4-Methoxybenzyl-acetat

Lieferant: Apollo Scientific

OR350225-100G
APOSOR350225-100GEA 0 EUR
APOSOR350225-100G
4-Methoxybenzyl-acetat
4-Methoxybenzyl-acetat
Formel: C₁₀H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00038509
CAS-Nummer: 104-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR