N-Phenyl-m-toluidin

Lieferant: Apollo Scientific

OR350228-100G
APOSOR350228-100GEA 0 EUR
APOSOR350228-100G
N-Phenyl-m-toluidin
N-Phenyl-m-toluidin
Formel: C₁₃H₁₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD00008530
CAS-Nummer: 1205-64-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR