Methyl-6-brom-2-naphthoat

Lieferant: Apollo Scientific

Synonyme: Methyl 6-bromo-2-naphthoate

OR350229-100G
APOSOR350229-100GEA 241 EUR
APOSOR350229-100G
Methyl-6-brom-2-naphthoat
Methyl-6-brom-2-naphthoat
Formel: C₁₂H₉BrO₂
Molecular Weight: 265,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00100408
CAS-Nummer: 33626-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR