Methyl-2,5-dihydroxybenzoat
Lieferant: Apollo Scientific
Synonyme:
2,5-Dihydroxybenzoesäuremethylester, Gentisinsäuremethylester
OR350235-100G
APOSOR350235-100GEA
71.2
EUR
APOSOR350235-100G
Methyl-2,5-dihydroxybenzoat
Methyl-2,5-dihydroxybenzoat
Formel:
C₈H₈O₄ Molecular Weight: 168,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016464 CAS-Nummer: 2150-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...