Methyl-3,4-dihydroxybenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,4-Dihydroxybenzoesäuremethylester, Protocatechusäuremethylester
OR350236-100G
APOSOR350236-100GEA
65.4
EUR
APOSOR350236-100G
Methyl-3,4-dihydroxybenzoat
Methyl-3,4-dihydroxybenzoat
Formel:
C₈H₈O₄ Molecular Weight: 168,15 g/mol Schmelzpunkt: 134…138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016437 CAS-Nummer: 2150-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...