1-Methyl-2-piperidinmethanol

Lieferant: Apollo Scientific

OR350237-100G
APOSOR350237-100GEA 81.2 EUR
APOSOR350237-100G
1-Methyl-2-piperidinmethanol
1-Methyl-2-piperidinmethanol
Formel: C₇H₁₅NO
Molecular Weight: 129,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006494
CAS-Nummer: 20845-34-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR