Methyl-5-formylsalicylat

Lieferant: Apollo Scientific

Synonyme: Methyl 5-Formyl-2-hydroxybenzoate

OR350239-100G
APOSOR350239-100GEA 243 EUR
APOSOR350239-100G
Methyl-5-formylsalicylat
Methyl-5-formylsalicylat
Formel: C₉H₈O₄
Molecular Weight: 180,1574 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840784
CAS-Nummer: 41489-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR