N-Phenyl-p-toluidin

Lieferant: Apollo Scientific

OR350243-25G
APOSOR350243-25GEA 0 EUR
APOSOR350243-25G
N-Phenyl-p-toluidin
N-Phenyl-p-toluidin
Formel: C₁₃H₁₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD00092921
CAS-Nummer: 620-84-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR